Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029746.3 | NP_084022.2 | 1, 2 | 193 | ||
GenBank mRNA | AK012236.1 | - | 1, 2 | 127 | ||
AK089920.1 | - | 1, 2 | 177 | |||
AK157348.1 | - | 1, 2 | 155 | |||
AK169298.1 | - | 1, 2 | 163 | |||
AK171726.1 | - | 1, 2 | 112 | |||
BC011435.1 | - | 1, 2 | 133 | 4010102 | ||
CJ093818.1 | - | 1, 2 | 193 |