Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029844.3 | NP_084120.1 | 3 | 829 | ||
GenBank mRNA | AK003912.2 | - | 3 | 830 | ||
AK158113.1 | - | 3 | 703 | |||
AY079153.1 | - | 3 | 606 | |||
BC027543.1 | - | 3 | 683 | 1477097 |