Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs7bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029879.2 | NP_084155.2 | 4 | 1234 | ||
GenBank mRNA | AK020910.1 | - | 4 | 831 | ||
AK028323.1 | - | 4 | 611 | |||
AK032138.1 | - | 4 | 1234 | |||
AK036240.1 | - | 4 | 1141 | |||
AK043529.1 | - | 1 | 38 | |||
AK046733.1 | - | 4 | 863 | |||
AK137417.1 | - | 4 | 1108 | |||
BC118957.2 | - | 4 | 766 | 40093423 | ||
DQ104214.1 | - | 4 | 524 |