Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dapk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285917.1 | NP_001272846.1 | 3 | 525 | ||
NM_029653.3 | NP_083929.2 | 3 | 525 | |||
NM_134062.2 | NP_598823.1 | 3 | 410 | |||
GenBank mRNA | AK009701.1 | - | 3 | 415 | ||
AK145922.1 | - | 3 | 411 | |||
AY245540.2 | - | 3 | 518 | |||
AY245541.1 | - | 3 | 511 | |||
BC057317.1 | - | 3 | 430 | 6821014 | ||
BC060161.1 | - | 3 | 430 | 6821014 | ||
X97048.1 | - | 2 | 360 |
Set Membership: |
![]() |