Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc35e3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029875.2 | NP_084151.2 | 5 | 3449 | ||
GenBank mRNA | AK020379.1 | - | 1 | 967 | ||
BC057101.1 | - | 5 | 3449 | 5720169 |