Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mboat7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029934.3 | NP_084210.2 | 8 | 2197 | ||
GenBank mRNA | AB055410.1 | - | 8 | 2195 | ||
AK019981.1 | - | 8 | 1608 | |||
AK150010.1 | - | 8 | 1591 | |||
AK154606.1 | - | 8 | 2196 | |||
AK158998.1 | - | 6 | 3118 | |||
AK170124.1 | - | 8 | 1674 | |||
BC023417.1 | - | 8 | 1645 | 5028759 | ||
BC118958.2 | - | 8 | 1842 | 40093821 | ||
EU016380.1 | - | 7 | 1409 |