Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TGFBI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000358.2 | NP_000349.1 | 2 | 381 | ||
GenBank mRNA | AK093916.1 | - | 2 | 236 | ||
AK094055.1 | - | 3 | 251 | |||
AK094581.1 | - | 2 | 325 | |||
AK222833.1 | - | 2 | 289 | |||
AK303643.1 | - | 2 | 285 | |||
BC000097.2 | - | 2 | 274 | 3506051 | ||
BC004972.1 | - | 2 | 269 | 2958878 | ||
BT009820.1 | - | 2 | 220 | |||
DA051240.1 | - | 2 | 381 | |||
KU178357.1 | - | 2 | 220 | |||
KU178358.1 | - | 2 | 220 | |||
M77349.1 | - | 2 | 267 |