Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164609.1 | NP_001158081.1 | 6 | 491 | ||
NM_001164610.1 | NP_001158082.1 | 6 | 491 | |||
NM_001164611.1 | NP_001158083.1 | 6 | 491 | |||
NM_029945.3 | NP_084221.2 | 6 | 491 | |||
GenBank mRNA | AK019466.2 | - | 6 | 492 | ||
AK031029.1 | - | 6 | 485 | |||
AK031685.1 | - | 6 | 490 | |||
AK035892.1 | - | 6 | 483 | |||
AK036055.1 | - | 6 | 483 | |||
AK036380.1 | - | 6 | 487 | |||
AK129354.1 | - | 6 | 468 | |||
AK138720.1 | - | 6 | 465 | |||
AK164234.1 | - | 6 | 483 | |||
AK164456.1 | - | 4 | 956 | |||
AK217585.1 | - | 3 | 147 | |||
BC026767.1 | - | 6 | 464 | 5137008 |