Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030127.3 | NP_084403.2 | 9 | 1775 | ||
GenBank mRNA | AK035194.1 | - | 9 | 1778 | ||
AY037300.1 | - | 9 | 1563 | |||
AY156509.1 | - | 9 | 1467 | |||
BC138587.1 | - | 9 | 1499 | 8861607 | ||
BC138588.1 | - | 9 | 1499 | 8861608 |