Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync1h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030238.2 | NP_084514.2 | 2 | 494 | ||
GenBank mRNA | AY004877.1 | - | 2 | 448 |