Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cttnbp2nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163332.1 | NP_001156804.1 | 5 | 579 | ||
NM_001163333.1 | NP_001156805.1 | 5 | 571 | |||
NM_030249.4 | NP_084525.1 | 5 | 687 | |||
GenBank mRNA | AK029901.1 | - | 5 | 688 | ||
AK030438.1 | - | 5 | 552 | |||
AK051259.1 | - | 4 | 3689 | |||
AK129359.1 | - | 5 | 577 | |||
AK145085.1 | - | 5 | 550 | |||
BC003236.1 | - | 5 | 541 | 3498078 | ||
BC006952.1 | - | 1 | 29 | |||
CV562515.1 | - | 5 | 571 | |||
EF410090.1 | - | 548 |