Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030611.3 | NP_085114.1 | 2 | 233 | ||
GenBank mRNA | AK149458.1 | - | 2 | 234 | ||
BC056643.1 | - | 2 | 215 | 5098276 | ||
D45850.1 | - | 2 | 185 |