Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038653.1 | NP_001033742.1 | 5 | 982 | ||
NM_001038654.1 | NP_001033743.1 | 4 | 887 | |||
NM_030696.3 | NP_109621.1 | 5 | 994 | |||
GenBank mRNA | AF178954.1 | - | 3 | 782 | ||
AK141381.1 | - | 4 | 886 | |||
AK149707.1 | - | 5 | 985 | |||
AK149983.1 | - | 5 | 985 | |||
AK165596.1 | - | 4 | 830 | |||
AK166048.1 | - | 970 | ||||
AK169917.1 | - | 5 | 944 | |||
AK171260.1 | - | 5 | 970 | |||
BC046525.1 | - | 5 | 994 | 5319754 |