Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031164.2 | NP_112441.2 | 2, 3 | 307 | ||
GenBank mRNA | AI115049.1 | - | 2, 3 | 297 | ||
AI527143.1 | - | 2, 3 | 303 | |||
AK149397.1 | - | 2, 3 | 294 | |||
AW213546.1 | - | 2, 3 | 307 | |||
BC030166.1 | - | 2, 3 | 292 | 5051604 | ||
D10071.1 | - | 2, 3 | 287 |