Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110101.2 | NP_001103571.1 | 2 | 276 | ||
NM_001204273.1 | NP_001191202.1 | 2 | 282 | |||
NM_001204274.1 | NP_001191203.1 | 2 | 276 | |||
NM_030597.3 | NP_085100.1 | 2 | 282 | |||
GenBank mRNA | AF204146.1 | - | 2 | 253 | ||
AF204147.1 | - | 2 | 253 | |||
AF204148.1 | - | 2 | 253 | |||
AF204149.1 | - | 2 | 253 | |||
AF204150.1 | - | 2 | 253 | |||
AF204151.1 | - | 2 | 253 | |||
AF204152.1 | - | 2 | 253 | |||
AF204153.1 | - | 1 | 495 | |||
AF204154.1 | - | 2 | 253 | |||
AF204155.1 | - | 2 | 253 | |||
AF204156.1 | - | 2 | 203 | |||
AF204157.1 | - | 2 | 120 | |||
AK151605.1 | - | 2 | 250 | |||
AK162880.1 | - | 2 | 268 | |||
AK178855.1 | - | 2 | 64 | |||
BB848823.1 | - | 2 | 282 | |||
BC014288.1 | - | 2 | 261 | 3982410 | ||
BC049543.1 | - | 275 | 6774564 |