Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030697.2 | NP_109622.1 | 11 | 2435 | ||
GenBank mRNA | AK002623.1 | - | 11 | 2436 | ||
AK132011.1 | - | 11 | 2457 | |||
BC037741.1 | - | 6 | 569 | |||
BC061055.1 | - | 6 | 581 | |||
BC115933.1 | - | 5 | 539 | |||
KJ650203.1 | - | 10 | 2366 | |||
KJ663713.1 | - | 10 | 2207 | |||
KJ663714.1 | - | 11 | 2228 |