Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Id4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031166.2 | NP_112443.1 | 3 | 631 | ||
GenBank mRNA | AK041164.1 | - | 3 | 830 | ||
BC145367.1 | - | 4 | 553 | |||
X75018.1 | - | 3 | 631 |