Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLEC SIRNA Assays ›
Gene Symbol:
PLEC
Gene Name:
plectin
Gene Aliases:
EBS1, EBSMD, EBSND, EBSO, EBSOG, EBSPA, HD1, LGMD2Q, PCN, PLEC1, PLEC1b, PLTN
Chromosome Location:
Chr.8: 143915147 - 143976800 on Build GRCh38
UniGene ID:
Hs.434248
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
8490
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLEC
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000445.4
NP_000436.2
5
419
NM_201378.3
NP_958780.1
4
304
NM_201379.2
NP_958781.1
4
372
NM_201380.3
NP_958782.1
4
870
NM_201381.2
NP_958783.1
4
267
NM_201382.3
NP_958784.1
4
357
NM_201383.2
NP_958785.1
4
310
NM_201384.2
NP_958786.1
4
418
XM_005250976.3
XP_005251033.1
5
912
XM_005250978.3
XP_005251035.1
5
437
XM_005250979.4
XP_005251036.1
5
396
XM_005250980.4
XP_005251037.1
5
481
XM_005250981.3
XP_005251038.1
5
318
XM_005250982.3
XP_005251039.1
5
311
XM_005250983.2
XP_005251040.1
4
235
XM_005250984.4
XP_005251041.1
5
300
XM_006716588.3
XP_006716651.1
6
489
XM_006716589.2
XP_006716652.1
5
250
XM_006716590.3
XP_006716653.1
5
360
XM_011517130.2
XP_011515432.1
5
437
XM_011517131.2
XP_011515433.1
4
344
XM_011517132.2
XP_011515434.1
5
433
GenBank mRNA
AY480044.1
-
4
376
AY480045.1
-
4
247
AY480046.1
-
4
372
AY480047.1
-
4
870
AY480048.1
-
4
267
AY480049.1
-
4
296
AY480050.1
-
4
310
AY480051.1
-
4
418
BP231761.1
-
4
357
BP312608.1
-
5
419
BP315878.1
-
4
305
BP359323.1
-
5
419
U53204.1
-
5
421
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001164303, NM_001164305, NM_001164296, NM_001164297, NM_001164308, and others.
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
hemidesmosome assembly
cell-cell adhesion
actin binding
protein binding
structural constituent of muscle
ankyrin binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0