Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tas1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031867.2 | NP_114073.1 | 1 | 189 | ||
GenBank mRNA | AF301161.1 | - | 1 | 85 | ||
AF301162.1 | - | 1 | 85 | |||
AF337040.1 | - | 1 | 267 | |||
AK153579.1 | - | 1 | 139 | |||
AY032622.1 | - | 1 | 85 | |||
BC070448.1 | - | 1 | 189 |
Set Membership: |
G-protein coupled receptor |