Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsph6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159671.1 | NP_001153143.1 | 1 | 298 | ||
NM_031255.2 | NP_112545.2 | 1 | 298 | |||
GenBank mRNA | AF329191.1 | - | 1 | 286 | ||
AF329192.1 | - | 1 | 291 | |||
AK029699.1 | - | 1 | 298 | |||
BC137992.1 | - | 1 | 291 | 8861011 |