Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Siglece | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031181.2 | NP_112458.2 | 6, 7 | 1222 | ||
GenBank mRNA | AF329269.1 | - | 6, 7 | 1226 | ||
AY348552.1 | - | 6, 7 | 1231 | |||
AY371487.1 | - | 6, 7 | 1211 | |||
BC023280.1 | - | 6, 7 | 1221 | 3662856 |