Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031251.4 | NP_112541.1 | 11 | 2452 | ||
GenBank mRNA | AJ250670.1 | - | 10 | 2350 | ||
AK078511.1 | - | 11 | 2464 | |||
AK079295.1 | - | 11 | 2453 | |||
AK137143.1 | - | 11 | 2495 | |||
AK170790.1 | - | 11 | 2464 | |||
AK170828.1 | - | 11 | 2475 | |||
BC005479.1 | - | 11 | 2439 | 3484952 | ||
BC006033.1 | - | 11 | 2439 |