Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040085.2 | NP_001035174.1 | 19 | 4412 | ||
NM_001040087.2 | NP_001035176.1 | 10 | 2619 | |||
NM_001040088.2 | NP_001035177.1 | 10 | 2547 | |||
NM_001289583.1 | NP_001276512.1 | 20 | 4460 | |||
NM_001289584.1 | NP_001276513.1 | 20 | 4379 | |||
NM_001289585.1 | NP_001276514.1 | 11 | 2712 | |||
NM_001289586.1 | NP_001276515.1 | 11 | 2667 | |||
NM_031394.3 | NP_113571.2 | 19 | 4340 | |||
GenBank mRNA | AB057754.1 | - | 19 | 3799 | ||
AB057755.1 | - | 13 | 4309 | |||
AB057756.1 | - | 11 | 2634 | |||
AB057757.1 | - | 6 | 2625 | |||
AB057760.1 | - | 19 | 3718 | |||
AB057761.1 | - | 18 | 3751 | |||
AB057762.1 | - | 18 | 3679 | |||
AB057763.1 | - | 7 | 2528 | |||
AK027924.1 | - | 10 | 2530 | |||
AK051653.1 | - | 11 | 2713 | |||
AK173212.1 | - | 10 | 2077 | |||
BC060665.1 | - | 11 | 4162 |