Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031842.2 | NP_114030.2 | 13 | 2361 | ||
GenBank mRNA | AK075611.1 | - | 9 | 1696 | ||
BC026783.1 | - | 13 | 2273 | 4481097 | ||
BC059921.1 | - | 13 | 2331 | |||
BC079839.1 | - | 13 | 2307 | 6810462 | ||
M25773.1 | - | 13 | 2310 | |||
U66620.1 | - | 13 | 2312 |