Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tas1r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031873.1 | NP_114079.1 | 4 | 1472 | ||
GenBank mRNA | AF337041.1 | - | 4 | 1472 | ||
AY032623.1 | - | 4 | 1441 | |||
BC137740.1 | - | 5 | 1374 | 8860759 | ||
BC144879.1 | - | 5 | 1374 | 9053427 |
Set Membership: |
![]() |