Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190490.1 | NP_001177419.1 | 2 | 333 | ||
NM_001190491.1 | NP_001177420.1 | 2 | 333 | |||
NM_032418.2 | NP_115794.1 | 2 | 333 | |||
GenBank mRNA | BC024150.1 | - | 2 | 320 | 4217643 | |
BC056615.1 | - | 2 | 312 | |||
BC075715.1 | - | 2 | 241 | 4989259 |