Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128597.1 | NP_001122069.1 | 2 | 569 | ||
NM_032460.2 | NP_115849.2 | 1 | 367 | |||
GenBank mRNA | AB221568.1 | - | 1 | 334 | ||
BC057902.1 | - | 2 | 528 | 5027029 | ||
BC120637.1 | - | 1 | 368 | 40129560 | ||
BC120639.1 | - | 1 | 368 | 40129562 |