Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: M1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033079.2 | NP_149070.2 | 2 | 165 | ||
GenBank mRNA | AF084364.1 | - | 1 | 134 | ||
AK131954.1 | - | 2 | 165 | |||
BC119803.1 | - | 1 | 92 | 40107692 | ||
BC122521.1 | - | 1 | 97 | 40107688 |