Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spred1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277256.1 | NP_001264185.1 | 3 | 914 | ||
NM_033524.3 | NP_277059.1 | 3 | 914 | |||
GenBank mRNA | AB063495.1 | - | 3 | 260 | ||
AK155065.1 | - | 3 | 877 | |||
AK155070.1 | - | 3 | 916 | |||
BC041685.1 | - | 2 | 228 | |||
BC057874.1 | - | 3 | 347 | 5319749 | ||
BC079606.1 | - | 3 | 535 | 30355455 |