Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032465.2 | NP_115854.2 | 2 | 344 | ||
GenBank mRNA | AK040329.1 | - | 2 | 231 | ||
AK040539.1 | - | 2 | 348 | |||
AK154076.1 | - | 2 | 314 | |||
AK156470.1 | - | 2 | 345 | |||
AY029156.1 | - | 2 | 205 | |||
BC052865.1 | - | 2 | 241 | 30072864 |