Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159986.1 | NP_001153458.1 | 2 | 376 | ||
NM_033354.3 | NP_203505.3 | 4 | 623 | |||
GenBank mRNA | AB063356.1 | - | 2 | 376 | ||
AK132144.1 | - | 2 | 400 | |||
AK173302.1 | - | 4 | 604 | |||
BC059194.1 | - | 3 | 441 | 4222915 |