Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289603.1 | NP_001276532.1 | 5, 6 | 585 | ||
NM_033476.3 | NP_258437.1 | 5, 6 | 585 | |||
GenBank mRNA | AK131989.1 | - | 4, 5 | 784 | ||
AK146503.1 | - | 5, 6 | 586 | |||
AK147898.1 | - | 4, 5 | 794 | |||
AK158682.1 | - | 5, 6 | 505 | |||
BC022131.1 | - | 5, 6 | 558 | 5098336 | ||
M84809.1 | - | 5, 6 | 524 |