Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TESK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320800.1 | NP_001307729.1 | 6, 7 | 811 | ||
NM_007170.2 | NP_009101.2 | 7, 8 | 1119 | |||
XM_005270355.3 | XP_005270412.1 | 6, 7 | 944 | |||
XM_006710287.2 | XP_006710350.1 | 8, 9 | 1159 | |||
XM_011540497.1 | XP_011538799.1 | 9, 10 | 1192 | |||
XM_011540500.2 | XP_011538802.1 | 7, 8 | 1091 | |||
XM_017000047.1 | XP_016855536.1 | 7, 8 | 1177 | |||
XM_017000048.1 | XP_016855537.1 | 8, 9 | 1447 | |||
XM_017000049.1 | XP_016855538.1 | 6, 7 | 1244 | |||
GenBank mRNA | AB057597.1 | - | 7, 8 | 1111 | ||
AJ132545.1 | - | 6, 7 | 1063 | |||
AK027573.1 | - | 7, 8 | 1093 | |||
AK294172.1 | - | 6, 7 | 786 | |||
BC022194.1 | - | 7, 8 | 1099 | |||
BC033085.1 | - | 6, 7 | 730 | 5425494 |