Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbscr16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033572.2 | NP_291050.1 | 2 | 489 | ||
GenBank mRNA | AF410456.1 | - | 2 | 468 | ||
AK005040.1 | - | 2 | 435 | |||
AK017733.1 | - | 2 | 488 | |||
BC024714.1 | - | 2 | 461 | 5134834 | ||
BC055335.1 | - | 2 | 487 | 6412407 |