Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033561.1 | NP_291039.1 | 4 | 389 | ||
GenBank mRNA | AK045004.1 | - | 4 | 391 | ||
AK088818.1 | - | 4 | 401 | |||
AK122194.1 | - | 4 | 389 | |||
AK132034.1 | - | 4 | 409 | |||
AK135390.1 | - | 4 | 390 | |||
AK136999.1 | - | 4 | 376 | |||
AK149841.1 | - | 4 | 375 | |||
AK150088.1 | - | 4 | 398 | |||
AK166567.1 | - | 4 | 390 | |||
AK168872.1 | - | 4 | 408 | |||
AK169312.1 | - | 4 | 372 | |||
AK169353.1 | - | 4 | 390 | |||
AK169407.1 | - | 4 | 408 | |||
AK178505.1 | - | 334 | ||||
AK182256.1 | - | 1 | 36 | |||
AK183328.1 | - | 3 | 150 | |||
AK184202.1 | - | 1 | 36 | |||
AK188319.1 | - | 1 | 45 | |||
AK191530.1 | - | 1 | 36 | |||
AK192457.1 | - | 1 | 36 | |||
AK193051.1 | - | 1 | 36 | |||
AK193532.1 | - | 1 | 36 | |||
AK193739.1 | - | 1 | 35 | |||
AK195570.1 | - | 1 | 36 | |||
AK196800.1 | - | 1 | 36 | |||
AK197150.1 | - | 222 | ||||
AK197761.1 | - | 1 | 36 | |||
AK200353.1 | - | 1 | 36 | |||
AK200533.1 | - | 1 | 36 | |||
AK201461.1 | - | 1 | 36 | |||
AK203547.1 | - | 1 | 36 | |||
AK213570.1 | - | 1 | 36 | |||
BC014796.1 | - | 4 | 403 | 3962104 |