Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dqx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033606.3 | NP_291084.1 | 3 | 659 | ||
GenBank mRNA | AF318278.1 | - | 2 | 303 | ||
AK010062.1 | - | 3 | 351 | |||
BC075680.1 | - | 3 | 659 | 30612145 |