Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198914.1 | NP_001185843.1 | 5 | 644 | ||
NM_010848.3 | NP_034978.3 | 5 | 644 | |||
GenBank mRNA | AK088079.1 | - | 5 | 565 | ||
AK088598.1 | - | 5 | 566 | |||
AK132250.1 | - | 5 | 509 | |||
AK137965.1 | - | 5 | 510 | |||
BC011513.1 | - | 5 | 552 | 3672769 | ||
M12848.1 | - | 5 | 642 | |||
X02774.1 | - | 5 | 414 |