Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Igsf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145800.1 | NP_001139272.1 | 21 | 4116 | ||
NM_033608.3 | NP_291086.2 | 21 | 4182 | |||
GenBank mRNA | AF317839.1 | - | 21 | 4169 | ||
AK078536.1 | - | 21 | 4112 | |||
AK145121.1 | - | 21 | 4113 | |||
BC023011.1 | - | 3 | 976 | |||
BC024487.1 | - | 4 | 941 | |||
BC034594.1 | - | 20 | 3945 | |||
BC040281.1 | - | 21 | 4142 | 5364073 |