Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163529.1 | NP_001157001.1 | 20 | 2766 | ||
NM_033615.2 | NP_291093.2 | 21 | 2844 | |||
GenBank mRNA | AB059632.1 | - | 21 | 2844 | ||
AB059633.1 | - | 20 | 2766 | |||
AF472524.1 | - | 21 | 2576 | |||
AK048367.1 | - | 16 | 2200 | |||
AK053825.1 | - | 17 | 3264 | |||
AK138517.1 | - | 4 | 856 | |||
AY382193.1 | - | 20 | 2484 | |||
AY382194.1 | - | 22 | 2489 | |||
AY953137.1 | - | 13 | 1787 |