Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pcdhb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053129.3 | NP_444359.3 | 1 | 1119 | ||
GenBank mRNA | AK029277.1 | - | 1 | 1111 | ||
AK036169.1 | - | 1 | 1120 | |||
AK143280.1 | - | 1 | 1120 | |||
AK160730.1 | - | 1 | 1120 | |||
AY013786.1 | - | 1 | 935 | |||
BC053098.1 | - | 1 | 1092 | 6401988 |