Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cadm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053199.3 | NP_444429.1 | 4 | 638 | ||
GenBank mRNA | AF195662.1 | - | 4 | 582 | ||
AK036352.1 | - | 5 | 694 | |||
AK038917.1 | - | 4 | 595 | |||
AK053077.1 | - | 4 | 592 | |||
AY059393.1 | - | 4 | 437 | |||
BC029659.1 | - | 1 | 57 | |||
BC079837.1 | - | 5 | 678 | |||
BC115417.2 | - | 4 | 543 | 40093410 | ||
BC115418.2 | - | 4 | 543 | 40093417 |