Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyve1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053247.4 | NP_444477.2 | 2 | 309 | ||
GenBank mRNA | AJ311501.1 | - | 2 | 464 | ||
AK004726.2 | - | 2 | 309 | |||
AK160857.1 | - | 1 | 47 | |||
BC038653.1 | - | 2 | 415 | 2936961 | ||
BC038892.1 | - | 2 | 415 | 3372151 |