Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053169.2 | NP_444399.2 | 6 | 1717 | ||
GenBank mRNA | AF220134.1 | - | 2 | 435 | ||
AF449496.1 | - | 6 | 1468 | |||
AK053139.1 | - | 6 | 1531 | |||
AK078902.1 | - | 4 | 1053 | |||
BC052821.1 | - | 7 | 1419 | 30057830 |