Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cadm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053199.3 | NP_444429.1 | 9 | 1957 | ||
GenBank mRNA | AF195662.1 | - | 9 | 1901 | ||
AK030782.1 | - | 4 | 1027 | |||
AK031927.1 | - | 1 | 364 | |||
AK053077.1 | - | 9 | 1911 | |||
AY059393.1 | - | 9 | 1755 | |||
BC029659.1 | - | 6 | 1376 | |||
BC079837.1 | - | 10 | 2397 | |||
BC115417.2 | - | 10 | 1382 | 40093410 | ||
BC115418.2 | - | 8 | 1393 | 40093417 |