Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Impa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053261.2 | NP_444491.1 | 8 | 1311 | ||
GenBank mRNA | AF353730.1 | - | 8 | 1310 | ||
AK080544.1 | - | 221 | ||||
AK154489.1 | - | 8 | 1349 | |||
AK154845.1 | - | 8 | 1282 | |||
AK170962.1 | - | 8 | 1349 | |||
AK171626.1 | - | 8 | 1284 | |||
BC011093.1 | - | 8 | 1286 | 4012163 |