Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080287.2 | NP_525026.2 | 19 | 1926 | ||
NM_207706.1 | NP_997589.1 | 20 | 1962 | |||
GenBank mRNA | AB093301.1 | - | 9 | 3197 | ||
AF398884.1 | - | 18 | 1825 | |||
AK032033.1 | - | 20 | 1935 | |||
AK033918.1 | - | 19 | 2120 | |||
AK035710.1 | - | 4 | 312 | |||
AK038097.1 | - | 19 | 1915 | |||
AK038455.1 | - | 19 | 2032 | |||
AK045428.1 | - | 19 | 1927 | |||
AK047040.1 | - | 19 | 1888 | |||
AK050823.1 | - | 19 | 1889 | |||
AK053574.1 | - | 20 | 1926 | |||
AK129023.1 | - | 19 | 1926 | |||
AK205725.1 | - | 2 | 119 | |||
BC006678.1 | - | 5 | 517 | |||
BC023954.1 | - | 20 | 1947 | 5320429 | ||
BC033418.1 | - | 18 | 1856 |