Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fezf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080433.3 | NP_536681.2 | 5 | 1811 | ||
GenBank mRNA | AI325906.1 | - | 2 | 461 | ||
AK019996.1 | - | 5 | 1775 |