Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080444.4 | NP_536692.1 | 6 | 1555 | ||
GenBank mRNA | AF398971.1 | - | 6 | 1584 | ||
AK028538.1 | - | 6 | 1555 | |||
AK052929.1 | - | 6 | 1471 | |||
BC141311.1 | - | 6 | 1546 | 9055923 | ||
BC141313.1 | - | 6 | 1546 | 9055925 |