Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080728.2 | NP_542766.1 | 4 | 235 | ||
GenBank mRNA | AK040973.1 | - | 4 | 236 | ||
AK147031.1 | - | 4 | 236 | |||
AK157742.1 | - | 4 | 236 | |||
AK169198.1 | - | 4 | 235 | |||
AK169371.1 | - | 4 | 235 | |||
AY056464.1 | - | 4 | 235 | |||
BC121789.1 | - | 3 | 191 | 40099448 | ||
BC158018.1 | - | 312 | 9007324 | |||
BC158070.1 | - | 4 | 490 | 9007368 | ||
DQ231556.1 | - | 4 | 238 | |||
DQ231557.1 | - | 3 | 262 |