Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Kirrel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170985.1 | NP_001164456.1 | 6 | 991 | ||
NM_130867.3 | NP_570937.2 | 6 | 988 | |||
GenBank mRNA | AF480411.1 | - | 5 | 626 | ||
AK158381.1 | - | 6 | 945 | |||
AY017368.1 | - | 5 | 626 | |||
AY243095.1 | - | 5 | 626 | |||
BC023765.1 | - | 6 | 804 | 5343720 |